4-Bromo-2,3-difluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3913-1G D3913-5G
TCD3913-1GEA 26.86 CAD
TCD3913-1G TCD3913-5G
4-Bromo-2,3-difluorophenol ≥98.0% (by GC, titration analysis)
4-Bromo-2,3-difluorophenol

CAS Number: 144292-32-0
MDL Number: MFCD08061907
Molecular Formula: C6H3BrF2O
Molecular Weight: 208.99
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 55

Formula: C₆H₃BrF₂O
Melting Pt: 55 °C
MDL Number: MFCD08061907
CAS Number: 144292-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d52b5d5820e7e7499aca396f32b62f04d63c991&vid=0d52b5d5820e7e7499aca396f32b62f04d63c991&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR