1-Benzhydrylazetidin-3-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3907-25G D3907-5G
TCD3907-25GEA 320.74 CAD
TCD3907-25G TCD3907-5G
1-Benzhydrylazetidin-3-ol ≥98.0% (by GC, titration analysis)
1-Benzhydrylazetidin-3-ol

CAS Number: 18621-17-5
MDL Number: MFCD00205109
Molecular Formula: C16H17NO
Molecular Weight: 239.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113


Other Product Names / Synonyms: 1-(Diphenylmethyl)-3-azetidinol, 1-Benzhydryl-3-azetidinol

Formula: C₁₆H₁₇NO
Melting Pt: 113 °C
MDL Number: MFCD00205109
CAS Number: 18621-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef425fa13aeb6f45a9c61b181b6e4649542dd23a&vid=ef425fa13aeb6f45a9c61b181b6e4649542dd23a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR