1,4-Dibromo-2,5-diiodobenzene ≥96.0% (by GC)

Supplier: TCI America

D3904-25G D3904-5G
TCD3904-25GEA 723.64 CAD
TCD3904-25G TCD3904-5G
1,4-Dibromo-2,5-diiodobenzene ≥96.0% (by GC)
1,4-Dibromo-2,5-diiodobenzene

CAS Number: 63262-06-6
MDL Number: MFCD00082898
Molecular Formula: C6H2Br2I2
Molecular Weight: 487.70
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 167

Formula: C₆H₂Br₂I₂
Melting Pt: 167 °C
MDL Number: MFCD00082898
CAS Number: 63262-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=130d4c0b093439d2f65dbaab32daaeb3f69c2694&vid=130d4c0b093439d2f65dbaab32daaeb3f69c2694&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR