Dichloro(1,10-phenanthroline)copper(II) ≥98.0%

Supplier: TCI America

Synonyms: (1,10-Phenanthroline)copper(II) Dichloride, Dichloro(1,10-phenanthroline)copper

D3891-1G D3891-5G
TCD3891-1GEA 71.1 CAD
TCD3891-1G TCD3891-5G
Dichloro(1,10-phenanthroline)copper(II) ≥98.0%
Dichloro(1,10-phenanthroline)copper(II)

CAS Number: 14783-09-6
MDL Number: MFCD00134224
Molecular Formula: C12H8Cl2CuN2
Molecular Weight: 314.66
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: (1,10-Phenanthroline)copper(II) Dichloride

Formula: C₁₂H₈Cl₂CuN₂
MW: 314.66 g/mol
MDL Number: MFCD00134224
CAS Number: 14783-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de980b1d1f6eaf2aeaf048899301309781e9e33a&vid=de980b1d1f6eaf2aeaf048899301309781e9e33a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR