2,3-Difluoro-6-methoxybenzaldehyde ≥97.0% (by GC)
Supplier: TCI America
D3885-1G
D3885-5G
TCD3885-1GEA
183.44
CAD
TCD3885-1G
TCD3885-5G
2,3-Difluoro-6-methoxybenzaldehyde ≥97.0% (by GC)
2,3-Difluoro-6-methoxybenzaldehyde
CAS Number: 187543-87-9
MDL Number: MFCD02093968
Molecular Formula: C8H6F2O2
Molecular Weight: 172.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 58
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3-Difluoro-6-methoxybenzaldehyde
Formula:
C₈H₆F₂O₂ Melting Pt: 58 °C |
MDL Number:
MFCD02093968 CAS Number: 187543-87-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...