N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-12, SB-12) ≥98.0% (by total nitrogen basis) for biochemical research

Supplier: TCI America

D3860-25G D3860-5G
TCD3860-25GEA 203.82 CAD
TCD3860-25G TCD3860-5G
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-12, SB-12) ≥98.0% (by total nitrogen basis) for biochemical research
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-12, SB-12)

[for Biochemical Research]
CAS Number: 14933-08-5
MDL Number: MFCD00036909
Molecular Formula: C17H37NO3S
Molecular Weight: 335.55
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 244
Flash Point (°C): 110


Product Categories: Bioscience, Amphoteric Surfactants (for Biochemistry)
Other Product Names / Synonyms: 3-(Dodecyldimethylammonio)propane-1-sulfonate, Lauryldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt, Lauryl Sulfobetaine, 3-(Lauryldimethylammonio)propane-1-sulfonate

Formula: C₁₇H₃₇NO₃S
Melting Pt: 244 °C
Flash Pt: 110 °C
MDL Number: MFCD00036909
CAS Number: 14933-08-5
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1a415c2bb523885afb2ada57d89e8389718c7af&vid=271893afe4c3dd0a828b111d88f6210a5de0062e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR