2,3-Dihydrobenzo[b]furan-5-carbaldehyde ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 5-Coumarancarboxaldehyde, 2,3-Dihydrobenzofuran-5-carbaldehyde, 2,3-Dihydrobenzo[b]furan-5-carboxaldehyde, 2,3-Dihydrobenzofuran-5-carboxaldehyde

D3857-25G D3857-5G
TCD3857-25GEA 489.8 CAD
TCD3857-25G TCD3857-5G
2,3-Dihydrobenzo[b]furan-5-carbaldehyde ≥96.0% (by GC)
2,3-Dihydrobenzo[b]furan-5-carbaldehyde

CAS Number: 55745-70-5
MDL Number: MFCD00068058
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 110
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: 5-Coumarancarboxaldehyde

Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 96 °C
Density: 1.20 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00068058
CAS Number: 55745-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5b0c9b164811ef4e96ecf9327872f9c9b07c0b2&vid=a5b0c9b164811ef4e96ecf9327872f9c9b07c0b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR