1,1-Dimethylsemicarbazide ≥96.0% (by GC, titration analysis)

Supplier: TCI America

D3854-25G D3854-5G
TCD3854-25GEA 604.98 CAD
TCD3854-25G TCD3854-5G
1,1-Dimethylsemicarbazide ≥96.0% (by GC, titration analysis)
1,1-Dimethylsemicarbazide

CAS Number: 22718-49-6
Molecular Formula: C3H9N3O
Molecular Weight: 103.13
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 140
Storage Temperature: 0-10°C

Formula: C₃H₉N₃O
Melting Pt: 140 °C
CAS Number: 22718-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e74297523c37dbd3ed8c1ddd159b1e946aeca74a&vid=e74297523c37dbd3ed8c1ddd159b1e946aeca74a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR