2,8-Quinolinediol ≥98.0%

Supplier: TCI America

Synonyms: 2,8-Dihydroxyquinoline

D3819-25G D3819-5G
TCD3819-25GEA 319 CAD
TCD3819-25G TCD3819-5G
2,8-Quinolinediol ≥98.0%
2,8-Quinolinediol

CAS Number: 15450-76-7
MDL Number: MFCD00216696
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 290
Lambda max.: 335 nm (EtOH)


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 2,8-Quinolinediol, 8-Hydroxy-2-quinolinone

Formula: C₉H₇NO₂
Melting Pt: 290 °C
MDL Number: MFCD00216696
CAS Number: 15450-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50bc82510bd84caaf80a260d8689d55846b57fea&vid=c5c317cfa639934030cf925d3b0b6829da222a91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR