Diisononyl cyclohexanedicarboxylate (mixture of isomers)

Supplier: TCI America

Synonyms: Cyclohexanedicarboxylic Acid Diisononyl Ester

D3754-25ML D3754-500ML
TCD3754-25MLEA 26.86 CAD
TCD3754-25ML TCD3754-500ML
Diisononyl cyclohexanedicarboxylate (mixture of isomers)
Diisononyl cyclohexanedicarboxylate (mixture of isomers)

CAS Number: 508181-38-2
Molecular Formula: C26H48O4
Molecular Weight: 424.67
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Cyclohexanedicarboxylic Acid Diisononyl Ester

Formula: C₂₆H₄₈O₄
MW: 424.67 g/mol
Density: 0.94 (20 °C)
CAS Number: 508181-38-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abe022be8a67b0a4f215c515bb83aefa1aa1752b&vid=abe022be8a67b0a4f215c515bb83aefa1aa1752b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR