Dimethyl-trans-cyclohexane-1,4-dicarboxylate ≥96.0%

Supplier: TCI America

Synonyms: Dimethyl trans-1,4-cyclohexanedicarboxylate, Dimethyl trans-1,4-Cyclohexanedicarboxylate

D3723-1G D3723-5G
TCD3723-1GEA 47.87 CAD
TCD3723-1G TCD3723-5G
Dimethyl-trans-cyclohexane-1,4-dicarboxylate ≥96.0%
Dimethyl trans-cyclohexane-1,4-dicarboxylate

CAS Number: 3399-22-2
MDL Number: MFCD00063917
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 69


Other Product Names / Synonyms: trans-1,4-Cyclohexanedicarboxylic Acid Dimethyl Ester

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Boiling Pt: 140 °C
Melting Pt: 69 °C
MDL Number: MFCD00063917
CAS Number: 3399-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01a10965d7cf196720e6c1fdb16bc012bec5f827&vid=01a10965d7cf196720e6c1fdb16bc012bec5f827&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR