2,6-Dimethylbenzothiazole ≥98.0% (by GC)

Supplier: TCI America

D3674-25G D3674-5G
TCD3674-25GEA 603.56 CAD
TCD3674-25G TCD3674-5G
2,6-Dimethylbenzothiazole ≥98.0% (by GC)
2,6-Dimethylbenzothiazole

CAS Number: 2941-71-1
Molecular Formula: C9H9NS
Molecular Weight: 163.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Specific Gravity (20/20): 1.14

Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 132 °C
Density: 1.14 (20 °C)
CAS Number: 2941-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806f92d3183756dcdf7ff9456c155b8b0a2f8922&vid=806f92d3183756dcdf7ff9456c155b8b0a2f8922&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR