4,6-Diaminopyrimidine ≥98.0%
Supplier: TCI America
Synonyms:
4,6-Pyrimidinediamine
D3648-1G
D3648-5G
TCD3648-1GEA
113.76
CAD
TCD3648-1G
TCD3648-5G
4,6-Diaminopyrimidine ≥98.0%
4,6-Diaminopyrimidine
CAS Number: 2434-56-2
Molecular Formula: C4H6N4
Molecular Weight: 110.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 271
Other Product Names / Synonyms: 4,6-Pyrimidinediamine
Formula:
C₄H₆N₄ MW: 110.12 g/mol Melting Pt: 271 °C |
MDL Number:
MFCD00044572 CAS Number: 2434-56-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...