2,4-Dichloro-5-methylpyrimidine ≥98.0% (by GC)

Supplier: TCI America

D3613-1G D3613-25G D3613-5G
TCD3613-1GEA 37.92 CAD
TCD3613-1G TCD3613-25G TCD3613-5G
2,4-Dichloro-5-methylpyrimidine ≥98.0% (by GC)
2,4-Dichloro-5-methylpyrimidine

CAS Number: 1780-31-0
MDL Number: MFCD00023197
Molecular Formula: C5H4Cl2N2
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 109
Flash Point (°C): 113
Freezing point (°C): 23

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 109 °C
Flash Pt: 113 °C
MDL Number: MFCD00023197
CAS Number: 1780-31-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17b1833fe7179da6c04de6b7736474f0eeb3eba6&vid=17b1833fe7179da6c04de6b7736474f0eeb3eba6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR