7-Hydroxy-3,4-dihydroquinolin-2(1H)-one ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D3599-25G D3599-5G
TCD3599-25GEA 69.52 CAD
TCD3599-25G TCD3599-5G
7-Hydroxy-3,4-dihydroquinolin-2(1H)-one ≥98.0% (by HPLC, total nitrogen)
7-Hydroxy-3,4-dihydroquinolin-2(1H)-one

CAS Number: 22246-18-0
MDL Number: MFCD06410891
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 236


Other Product Names / Synonyms: 7-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline

Formula: C₉H₉NO₂
Melting Pt: 236 °C
MDL Number: MFCD06410891
CAS Number: 22246-18-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afaeca90eca2ec249ee33de0d6588a631eb10d57&vid=afaeca90eca2ec249ee33de0d6588a631eb10d57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR