9,9-Diphenylfluorene ≥98.0% (by GC)

Supplier: TCI America

D3586-25G D3586-5G
TCD3586-25GEA 619.83 CAD
TCD3586-25G TCD3586-5G
9,9-Diphenylfluorene ≥98.0% (by GC)
9,9-Diphenylfluorene

CAS Number: 20302-14-1
MDL Number: MFCD00093944
Molecular Formula: C25H18
Molecular Weight: 318.42
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 226

Formula: C₂₅H₁₈
Melting Pt: 226 °C
MDL Number: MFCD00093944
CAS Number: 20302-14-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=086741e307f70041b0a18bed93415e046746b0be&vid=086741e307f70041b0a18bed93415e046746b0be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR