4,6-Dichloro-2-methylpyrimidine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Methyl-4,6-dichloropyrimidine
D3558-25G
D3558-5G
TCD3558-25GEA
191.18
CAD
TCD3558-25G
TCD3558-5G
4,6-Dichloro-2-methylpyrimidine ≥98.0% (by GC)
4,6-Dichloro-2-methylpyrimidine
CAS Number: 1780-26-3
MDL Number: MFCD00090472
Molecular Formula: C5H4Cl2N2
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 47
Flash Point (°C): 98
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol Melting Pt: 47 °C Flash Pt: 98 °C |
MDL Number:
MFCD00090472 CAS Number: 1780-26-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...