2,5-Dibromo-3-picoline ≥98.0%

Supplier: TCI America

Synonyms: 2,5-Dibromo-3-methylpyridine

D3455-25G D3455-5G
TCD3455-25GEA 79.32 CAD
TCD3455-25G TCD3455-5G
2,5-Dibromo-3-picoline ≥98.0%
2,5-Dibromo-3-picoline

CAS Number: 3430-18-0
MDL Number: MFCD02093085
Molecular Formula: C6H5Br2N
Molecular Weight: 250.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 46
Flash Point (°C): 110


Other Product Names / Synonyms: 2,5-Dibromo-3-picoline

Formula: C₆H₅Br₂N
MW: 250.92
Boiling Pt: 114 °C
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD02093085
CAS Number: 3430-18-0
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0f0e3cb947f6265c0d941334b3a1adfb939983&vid=5f0f0e3cb947f6265c0d941334b3a1adfb939983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR