4,4'-Dihydroxyazobenzene ≥98.0%

Supplier: TCI America

Synonyms: 4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol

D3431-1G D3431-5G
TCD3431-1GEA 304.94 CAD
TCD3431-1G TCD3431-5G
4,4'-Dihydroxyazobenzene ≥98.0%
4,4'-Dihydroxyazobenzene

CAS Number: 2050-16-0
MDL Number: MFCD00045778
Molecular Formula: C12H10N2O2
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 215
Lambda max.: 360 nm (EtOH)


Other Product Names / Synonyms: 4,4'-Azodiphenol

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Melting Pt: 215 °C
MDL Number: MFCD00045778
CAS Number: 2050-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fbec01160ce8bdf9bfb2bb4f02eefcaf2b40422&vid=0fbec01160ce8bdf9bfb2bb4f02eefcaf2b40422&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR