7-(Diethylamino)coumarin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3354-1G D3354-200MG
TCD3354-1GEA 281.24 CAD
TCD3354-1G TCD3354-200MG
7-(Diethylamino)coumarin ≥98.0% (by GC, titration analysis)
7-(Diethylamino)coumarin

CAS Number: 20571-42-0
MDL Number: MFCD00232946
Molecular Formula: C13H15NO2
Molecular Weight: 217.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 89
Lambda max.: 360 nm (Cyclohexane)

Formula: C₁₃H₁₅NO₂
Melting Pt: 89 °C
MDL Number: MFCD00232946
CAS Number: 20571-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5558704892ccc02094d13948819079de292b83d&vid=c5558704892ccc02094d13948819079de292b83d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR