3,4-Dihydroxybenzophenone ≥98.0%

Supplier: TCI America

Synonyms: (3,4-Dihydroxyphenyl)(phenyl)methanone

D3347-25G D3347-5G
TCD3347-25GEA 183.28 CAD
TCD3347-25G TCD3347-5G
3,4-Dihydroxybenzophenone ≥98.0%
3,4-Dihydroxybenzophenone

CAS Number: 10425-11-3
MDL Number: MFCD00477203
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 4-Benzoylpyrocatechol

Formula: C₁₃H₁₀O₃
Melting Pt: 148 °C
MDL Number: MFCD00477203
CAS Number: 10425-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e551fb6d88474c70b76a5ab2a7105152fea3013d&vid=e551fb6d88474c70b76a5ab2a7105152fea3013d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR