3,8-Dibromo-1,10-phenanthroline ≥96.0%

Supplier: TCI America

D3209-1G D3209-200MG
TCD3209-1GEA 396.74 CAD
TCD3209-1G TCD3209-200MG
3,8-Dibromo-1,10-phenanthroline ≥96.0%
3,8-Dibromo-1,10-phenanthroline

CAS Number: 100125-12-0
MDL Number: MFCD09909860
Molecular Formula: C12H6Br2N2
Molecular Weight: 338.00
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 282
Lambda max.: 352 nm (DMF)

Formula: C12H6Br2N2
MW: 338.0000 g/mol
Melting Pt: 282 °C
MDL Number: MFCD09909860
CAS Number: 100125-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=703e88341a19d1f161027ab7000ab69f872d6666&vid=703e88341a19d1f161027ab7000ab69f872d6666&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR