Diethyl (2-oxopropyl)phosphonate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Diethyl acetylmethylphosphonate

D3174-25G D3174-5G
TCD3174-25GEA 478.74 CAD
TCD3174-25G TCD3174-5G
Diethyl (2-oxopropyl)phosphonate ≥95.0% (by GC)
Diethyl (2-oxopropyl)phosphonate

CAS Number: 1067-71-6
MDL Number: MFCD00044728
Molecular Formula: C7H15O4P
Molecular Weight: 194.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 113
Specific Gravity (20/20): 1.11


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (2-Oxopropyl)phosphonic Acid Diethyl Ester, Diethyl Acetonylphosphonate, Acetonylphosphonic Acid Diethyl Ester

Formula: C₇H₁₅O₄P
MW: 194.17 g/mol
Boiling Pt: 126 °C
Density: 1.11 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00044728
CAS Number: 1067-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=018ba4b96676d416d0e29112648ed385bdd64518&vid=018ba4b96676d416d0e29112648ed385bdd64518&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR