1,6-Dibromopyrene ≥98.0%

Supplier: TCI America

D3167-1G D3167-5G
TCD3167-1GEA 51.19 CAD
TCD3167-1G TCD3167-5G
1,6-Dibromopyrene ≥98.0%
1,6-Dibromopyrene

CAS Number: 27973-29-1
MDL Number: MFCD09263746
Molecular Formula: C16H8Br2
Molecular Weight: 360.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 230
Lambda max.: 246 nm (EtOH)

Formula: C₁₆H₈Br₂
MW: 360.05 g/mol
Melting Pt: 230 °C
MDL Number: MFCD09263746
CAS Number: 27973-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c23724db43200d9d9436764bf57474e3b92b356&vid=6c23724db43200d9d9436764bf57474e3b92b356&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR