4,4'-Di-tert-butyl-2,2'-bipyridine ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Di-tert-butyl-2,2'-bipyridyl, 4,4'-Di-tert-butyl-2,2'-bipyridine
CAS Number: 72914-19-3
MDL Number: MFCD01863731
Molecular Formula: C18H24N2
Molecular Weight: 268.40
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 161
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: 4,4'-Di-tert-butyl-2,2'-bipyridine
Formula:
C18H24N2 MW: 268.4000 g/mol Melting Pt: 161 °C |
MDL Number:
MFCD01863731 CAS Number: 72914-19-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...