3,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
Dihydroxy(3,4-dimethylphenyl)borane, (3,4-Dimethylphenyl)metaboric acid, 3,4-Dimethylbenzeneboronic acid
D3110-1G
D3110-5G
TCD3110-1GEA
93.22
CAD
TCD3110-1G
TCD3110-5G
3,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
3,4-Dimethylphenylboronic acid
CAS Number: 55499-43-9
MDL Number: MFCD01009694
Molecular Formula: C8H11BO2
Molecular Weight: 149.98
Form: Crystal
Color: White
Other Product Names / Synonyms: 3,4-Dimethylbenzeneboronic Acid
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol |
MDL Number:
MFCD01009694 CAS Number: 55499-43-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...