2-Deoxy-2-fluoro-D-glucose ≥98.0%
Supplier: TCI America
D3023-100MG
TCD3023-100MGEA
185.18
CAD
TCD3023-100MG
2-Deoxy-2-fluoro-D-glucose ≥98.0%
2-Deoxy-2-fluoro-D-glucose
CAS Number: 29702-43-0
MDL Number: MFCD00077527
Molecular Formula: C6H11FO5
Molecular Weight: 182.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 176
Specific rotation [a]20/D: 63 deg (C=1, dilute NH3aq.)
Storage Temperature: <0°C
Product Categories: Bioscience, Glucose
Other Product Names / Synonyms: 2-Deoxy-2-fluoro-D-glucose
Formula:
C₆H₁₁FO₅ Melting Pt: 176 °C |
MDL Number:
MFCD00077527 CAS Number: 29702-43-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...