2,4-Dichloro-6-methylpyrimidine ≥98.0% (by GC)

Supplier: TCI America

D2958-25G D2958-5G
TCD2958-25GEA 290.88 CAD
TCD2958-25G TCD2958-5G
2,4-Dichloro-6-methylpyrimidine ≥98.0% (by GC)
2,4-Dichloro-6-methylpyrimidine

CAS Number: 5424-21-5
MDL Number: MFCD00006064
Molecular Formula: C5H4Cl2N2
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 219
Melting point (°C): 47
Flash Point (°C): 230

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 219 °C
Melting Pt: 47 °C
Flash Pt: 230 °C
MDL Number: MFCD00006064
CAS Number: 5424-21-5
UN: 3263
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=025e1b224015d739fa151ac4786eb4a0283c2e1a&vid=025e1b224015d739fa151ac4786eb4a0283c2e1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR