3,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2927-1G D2927-5G
TCD2927-1GEA 86.9 CAD
TCD2927-1G TCD2927-5G
3,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)
3,4-Dichlorophenethylamine

CAS Number: 21581-45-3
MDL Number: MFCD00060617
Molecular Formula: C8H9Cl2N
Molecular Weight: 190.07
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.27


Other Product Names / Synonyms: 1-Amino-2-(3,4-dichlorophenyl)ethane, 4-(2-Aminoethyl)-1,2-dichlorobenzene, 3,4-Dichlorophenethylamine

Formula: C₈H₉Cl₂N
MW: 190.07 g/mol
Density: 1.27 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00060617
CAS Number: 21581-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b3df4651233b2cee958d39e4b3ccab0fc1b761d&vid=8b3df4651233b2cee958d39e4b3ccab0fc1b761d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR