4,4'-Diamino-2,2'-bipyridyl ≥98.0%

Supplier: TCI America

Synonyms: [2,2'-Bipyridine]-4,4'-diamine, 4,4'-Diamino-2,2'-bipyridine

D2892-1G D2892-200MG
TCD2892-1GEA 337.52 CAD
TCD2892-1G TCD2892-200MG
4,4'-Diamino-2,2'-bipyridyl ≥98.0%
4,4'-Diamino-2,2'-bipyridyl

CAS Number: 18511-69-8
MDL Number: MFCD08704188
Molecular Formula: C10H10N4
Molecular Weight: 186.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 4,4'-Diamino-2,2'-bipyridine

Formula: C₁₀H₁₀N₄
MW: 186.22 g/mol
MDL Number: MFCD08704188
CAS Number: 18511-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99d6bc753154ed67b4c013b9e223dc4a91fae854&vid=99d6bc753154ed67b4c013b9e223dc4a91fae854&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR