Nickel diethyldithiocarbamate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D2879-25G D2879-500G
TCD2879-25GEA 53.72 CAD
TCD2879-25G TCD2879-500G
Nickel diethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Nickel diethyldithiocarbamate

CAS Number: 14267-17-5
MDL Number: MFCD00137706
Molecular Formula: C5H11NS2
Molecular Weight: 355.22
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Flash Point (°C): 267


Other Product Names / Synonyms: Diethyldithiocarbamic Acid Nickel Salt

Formula: C₅H₁₁NS₂
Flash Pt: 267 °C
MDL Number: MFCD00137706
CAS Number: 14267-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fd742f54ec4fb76840aec32e33025d0114c152e&vid=7fd742f54ec4fb76840aec32e33025d0114c152e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR