3,5-Dibromobenzaldehyde ≥97.0% (by GC)
Supplier: TCI America
D2832-25G
D2832-5G
TCD2832-25GEA
399.74
CAD
TCD2832-25G
TCD2832-5G
3,5-Dibromobenzaldehyde ≥97.0% (by GC)
3,5-Dibromobenzaldehyde
CAS Number: 56990-02-4
MDL Number: MFCD00156887
Molecular Formula: C7H4Br2O
Molecular Weight: 263.92
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 93
Formula:
C₇H₄Br₂O Melting Pt: 93 °C |
MDL Number:
MFCD00156887 CAS Number: 56990-02-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...