(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥98.0% (by GC)

Supplier: TCI America

D2774-100MG D2774-1G
TCD2774-100MGEA 229.1 CAD
TCD2774-100MG TCD2774-1G
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥98.0% (by GC)
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl

CAS Number: 145964-33-6
MDL Number: MFCD00269686
Molecular Formula: C33H25OP
Molecular Weight: 468.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 193
Specific rotation [a]20/D: 94 deg (C=0.3, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: (R)-(+)-MOP

Formula: C₃₃H₂₅OP
Melting Pt: 193 °C
MDL Number: MFCD00269686
CAS Number: 145964-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b5cdaf5616f3fbef22af726837db11c63a96dd&vid=28b5cdaf5616f3fbef22af726837db11c63a96dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR