3,5-Difluorophenol ≥98.0% (by GC)
Supplier: TCI America
D2569-25G
D2569-5G
TCD2569-25GEA
240.16
CAD
TCD2569-25G
TCD2569-5G
3,5-Difluorophenol ≥98.0% (by GC)
3,5-Difluorophenol
CAS Number: 2713-34-0
MDL Number: MFCD00002255
Molecular Formula: C6H4F2O
Molecular Weight: 130.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 68
Melting point (°C): 54
Flash Point (°C): 70
Formula:
C₆H₄F₂O Boiling Pt: 68 °C Melting Pt: 54 °C Flash Pt: 70 °C |
MDL Number:
MFCD00002255 CAS Number: 2713-34-0 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...