Diphenyl-1-pyrenylphosphine ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
DPPP, Diphenyl(pyren-1-yl)phosphine, 1-(Diphenylphosphino)pyrene
D2404-100MG
D2404-1G
TCD2404-100MGEA
192.87
CAD
TCD2404-100MG
TCD2404-1G
Diphenyl-1-pyrenylphosphine ≥95.0% (by GC)
Diphenyl-1-pyrenylphosphine
CAS Number: 110231-30-6
Molecular Formula: C28H19P
Molecular Weight: 386.43
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 176
Storage Temperature: 0-10°C
Product Categories: Bioscience, Fluorescent Probes, ROS/RNS Detection Reagents, Redox Research Reagents
Other Product Names / Synonyms: 1-(Diphenylphosphino)pyrene, DPPP
Formula:
C₂₈H₁₉P MW: 386.43 g/mol Melting Pt: 176 °C |
MDL Number:
MFCD00278755 CAS Number: 110231-30-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...