4,4'-Dibromotriphenylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1910-1G D1910-5G
TCD1910-1GEA 77.42 CAD
TCD1910-1G TCD1910-5G
4,4'-Dibromotriphenylamine ≥98.0% (by GC, titration analysis)
4,4'-Dibromotriphenylamine

CAS Number: 81090-53-1
MDL Number: MFCD00060104
Molecular Formula: C18H13Br2N
Molecular Weight: 403.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 85
Flash Point (°C): 110


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: N,N-Bis(p-bromophenyl)aniline

Formula: C₁₈H₁₃Br₂N
Melting Pt: 85 °C
Flash Pt: 110 °C
MDL Number: MFCD00060104
CAS Number: 81090-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecb8c47fc0bd9cbf5509b2e28958087f4d8685ad&vid=ecb8c47fc0bd9cbf5509b2e28958087f4d8685ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR