Sodium dichloroacetate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
D1719-10G
D1719-25G
TCD1719-10GEA
91.97
CAD
TCD1719-10G
TCD1719-25G
Sodium dichloroacetate ≥99.0% (by titrimetric analysis)
Sodium dichloroacetate
CAS Number: 2156-56-1
MDL Number: MFCD00070489
Molecular Formula: C2H2Cl2O2
Molecular Weight: 150.92
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Other Product Names / Synonyms: Dichloroacetic Acid Sodium Salt
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...