N,N-Dimethylformamide dineopentyl acetal ≥96.0% (by GC)

Supplier: TCI America

D1595-25ML D1595-5ML
TCD1595-25MLEA 200.66 CAD
TCD1595-25ML TCD1595-5ML
N,N-Dimethylformamide dineopentyl acetal ≥96.0% (by GC)
N,N-Dimethylformamide dineopentyl acetal

[for Esterification]
CAS Number: 4909-78-8
MDL Number: MFCD00008851
Molecular Formula: C13H29NO2
Molecular Weight: 231.38
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Flash Point (°C): 63
Specific Gravity (20/20): 0.83

Formula: C₁₃H₂₉NO₂
MW: 231.38 g/mol
Boiling Pt: 87 °C
Density: 0.83 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00008851
CAS Number: 4909-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f5c1893fe31704da52439d4d2ae806551960174&vid=1d1619c8898ee4a375d5e26ea3ae97c3fc66320e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR