Ethyl erucate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl docos-13-enoate, Erucic Acid Ethyl Ester, cis-13-Docosenoic Acid Ethyl Ester, Ethyl cis-13-Docosenoate

D1014-25ML D1014-5ML
TCD1014-25MLEA 386.78 CAD
TCD1014-25ML TCD1014-5ML
Ethyl erucate ≥95.0%
Ethyl erucate

CAS Number: 37910-77-3
MDL Number: MFCD00027405
Molecular Formula: C24H46O2
Molecular Weight: 366.63
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 233
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: cis-13-Docosenoic Acid Ethyl Ester, Ethyl cis-13-Docosenoate, Erucic Acid Ethyl Ester

Formula: C₂₄H₄₆O₂
Boiling Pt: 233 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00027405
CAS Number: 37910-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fac910710fd65e95fe9f779efd851b3bb672c05f&vid=fac910710fd65e95fe9f779efd851b3bb672c05f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR