1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98.0% stabilised

Supplier: TCI America

Synonyms: Diethylene glycol divinyl ether, 1,1'-[Oxybis(ethyleneoxy)]diethene, 1,1'-(Oxybis(ethyleneoxy))diethylene, 1,1'-(Oxybis(ethyleneoxy))diethene, Diethyleneglycoldivinylether, (2-(2-Ethenyloxyethoxy)ethoxy)ethene

D0498-100ML D0498-25ML
TCD0498-100MLEA 317.58 CAD
TCD0498-100ML TCD0498-25ML
1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98.0% stabilised
1,1'-[Oxybis(ethyleneoxy)]diethylene

(stabilized with KOH)
CAS Number: 764-99-8
MDL Number: MFCD00054610
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Flash Point (°C): 71
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Divinyl Carbitol

Formula: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MW: 158.2 g/mol
Boiling Pt: 197 °C
Density: 0.97 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00054610
CAS Number: 764-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd05326ae581d527555ae6447d2809345724a5dd&vid=bd05326ae581d527555ae6447d2809345724a5dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR