1,4-Diaminobutane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0239-25G D0239-400G
TCD0239-25GEA 53.96 CAD
TCD0239-25G TCD0239-400G
1,4-Diaminobutane ≥98.0% (by GC, titration analysis)
1,4-Diaminobutane

CAS Number: 110-60-1
MDL Number: MFCD00008235
Molecular Formula: C4H12N2
Molecular Weight: 88.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 159
Flash Point (°C): 60
Freezing point (°C): 27


Product Categories: Materials Chemistry, Monomers, Bioscience, Other Plant Hormones, Polyamines
Other Product Names / Synonyms: 1,4-Butanediamine, Putrescine, Tetramethylenediamine

Formula: C₄H₁₂N₂
Boiling Pt: 159 °C
Flash Pt: 60 °C
MDL Number: MFCD00008235
CAS Number: 110-60-1
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e65d4281d10d3dc19500b7fc97fb35051fdc899c&vid=e65d4281d10d3dc19500b7fc97fb35051fdc899c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR