1,3-Dithiane ≥97.0% (by GC)
Supplier: TCI America
D0119-25G
D0119-5G
TCD0119-25GEA
422.92
CAD
TCD0119-25G
TCD0119-5G
1,3-Dithiane ≥97.0% (by GC)
1,3-Dithiane
CAS Number: 505-23-7
MDL Number: MFCD00006654
Molecular Formula: C4H8S2
Molecular Weight: 120.23
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 208
Melting point (°C): 53
Formula:
C₄H₈S₂ Boiling Pt: 208 °C Melting Pt: 53 °C |
MDL Number:
MFCD00006654 CAS Number: 505-23-7 UN: 3335 ADR: 9, |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...