6-Chloro-2-pyridinecarboxaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 6-Chloropyridine-2-carboxaldehyde, 6-Chloro-2-formylpyridine, 6-Chloropicolinaldehyde

C3017-1G C3017-5G
TCC3017-1GEA 195.92 CAD
TCC3017-1G TCC3017-5G
6-Chloro-2-pyridinecarboxaldehyde ≥98.0%
6-Chloro-2-pyridinecarboxaldehyde

CAS Number: 54087-03-5
MDL Number: MFCD09832941
Molecular Formula: C6H4ClNO
Molecular Weight: 141.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 71


Other Product Names / Synonyms: 6-Chloropicolinaldehyde, 6-Chloro-2-formylpyridine

Formula: C₆H₄ClNO
MW: 141.55 g/mol
Melting Pt: 71 °C
MDL Number: MFCD09832941
CAS Number: 54087-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbd58cf6ac4728aa0282870f05aaa6d339cea21b&vid=d5a4ef6ed3bb87919e4174c92a38de13e3774388&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR