2-Chloro-N-methylacetamide ≥98.0%

Supplier: TCI America

C3014-25G C3014-5G
TCC3014-25GEA 334.96 CAD
TCC3014-25G TCC3014-5G
2-Chloro-N-methylacetamide ≥98.0%
2-Chloro-N-methylacetamide

CAS Number: 96-30-0
MDL Number: MFCD00018913
Molecular Formula: C3H6ClNO
Molecular Weight: 107.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 116
Melting point (°C): 44

Formula: C₃H₆ClNO
MW: 107.54 g/mol
Boiling Pt: 116 °C
Melting Pt: 44 °C
MDL Number: MFCD00018913
CAS Number: 96-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11c432c183b26e3793e9f3453a0f398ecef56c81&vid=11c432c183b26e3793e9f3453a0f398ecef56c81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR