3-Chloro-1,2,4-triazole ≥98.0%
Supplier: TCI America
C3013-1G
C3013-5G
TCC3013-1GEA
158.63
CAD
TCC3013-1G
TCC3013-5G
3-Chloro-1,2,4-triazole ≥98.0%
3-Chloro-1,2,4-triazole
CAS Number: 6818-99-1
MDL Number: MFCD00128812
Molecular Formula: C2H2ClN3
Molecular Weight: 103.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 169
Formula:
C₂H₂ClN₃ MW: 103.51 g/mol Melting Pt: 169 °C |
MDL Number:
MFCD00128812 CAS Number: 6818-99-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...