4-Chloro-5'-phenyl-1,1':3',1''-terphenyl ≥98.0%

Supplier: TCI America

C3008-1G C3008-200MG
TCC3008-1GEA 310.79 CAD
TCC3008-1G TCC3008-200MG
4-Chloro-5'-phenyl-1,1':3',1''-terphenyl ≥98.0%
4-Chloro-5'-phenyl-1,1':3',1''-terphenyl

CAS Number: 116941-51-6
Molecular Formula: C24H17Cl
Molecular Weight: 340.85
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 143
Lambda max.: 254 nm (CH3CN)


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 5'-(4-Chlorophenyl)-1,1':3',1''-terphenyl

Formula: C₂₄H₁₇Cl
Melting Pt: 143 °C
CAS Number: 116941-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e158a57331523bcd3cfd90a46e7e1905f247c41&vid=5e158a57331523bcd3cfd90a46e7e1905f247c41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR