4-Chloro-D-mandelic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid

C3006-1G C3006-5G
TCC3006-1GEA 64.78 CAD
TCC3006-1G TCC3006-5G
4-Chloro-D-mandelic acid ≥98.0% (by GC, titration analysis)
4-Chloro-D-mandelic acid

CAS Number: 32189-36-9
MDL Number: MFCD00798430
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 121
Specific rotation [a]20/D: -136 deg (C=0.3, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
Melting Pt: 121 °C
MDL Number: MFCD00798430
CAS Number: 32189-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df83233423e385aaf0699aff3719e4d5fcd2b341&vid=29a900a7a992edbb3921c56f02e74005d4e5f433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR