5-Chloro-2-formylpyridine ≥98.0% (by GC)

Supplier: TCI America

C3000-1G C3000-5G
TCC3000-1GEA 107.44 CAD
TCC3000-1G TCC3000-5G
5-Chloro-2-formylpyridine ≥98.0% (by GC)
5-Chloro-2-formylpyridine

CAS Number: 31181-89-2
MDL Number: MFCD08277271
Molecular Formula: C6H4ClNO
Molecular Weight: 141.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65


Other Product Names / Synonyms: 5-Chloro-2-formylpyridine, 5-Chloropicolinaldehyde

Formula: C₆H₄ClNO
Melting Pt: 65 °C
MDL Number: MFCD08277271
CAS Number: 31181-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c32db5b259c80c6db6502e29ebf5027eb112339e&vid=c32db5b259c80c6db6502e29ebf5027eb112339e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR