(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥98.0%

Supplier: TCI America

Synonyms: (S)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate, Benzyl (S)-Tetrahydro-5-oxo-3-furanylcarbamate, (S)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (S)-beta-(Cbz-amino)-gamma-butyrolactone, (S)-4-(Cbz-amino)tetrahydrofuran-2-one

C2995-1G C2995-5G
TCC2995-1GEA 58.46 CAD
TCC2995-1G TCC2995-5G
(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥98.0%
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone

CAS Number: 87219-29-2
MDL Number: MFCD00216569
Molecular Formula: C12H13NO4
Molecular Weight: 235.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 106
Specific rotation [a]20/D: -55 deg (C=1, CHCl3)


Other Product Names / Synonyms: (S)-beta-(Cbz-amino)-gamma-butyrolactone, (S)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (S)-4-(Cbz-amino)tetrahydrofuran-2-one, Benzyl (S)-Tetrahydro-5-oxo-3-furanylcarbamate

Formula: C₁₂H₁₃NO₄
Melting Pt: 106 °C
MDL Number: MFCD00216569
CAS Number: 87219-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0501b015e7607fcdff9d686e8940e7a7c298219c&vid=0501b015e7607fcdff9d686e8940e7a7c298219c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR