N-(3-Chlorophenyl)-3-oxobutyramide 96,0
Supplier: TCI America
Synonyms:
3'-Chloroacetoacetanilide
C2984-1G
C2984-200MG
TCC2984-1GEA
647.96
CAD
TCC2984-1G
TCC2984-200MG
N-(3-Chlorophenyl)-3-oxobutyramide 96,0
N-(3-Chlorophenyl)-3-oxobutyramide
CAS Number: 2415-87-4
MDL Number: MFCD00018440
Molecular Formula: C10H10ClNO2
Molecular Weight: 211.65
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 103
Lambda max.: 247 nm (EtOH)
Product Categories: Synthetic Organic Chemistry, Diketone Compounds (Multi-component Reaction)
Other Product Names / Synonyms: 3'-Chloroacetoacetanilide
Formula:
C₁₀H₁₀ClNO₂ MW: 211.65 g/mol Melting Pt: 103 °C |
MDL Number:
MFCD00018440 CAS Number: 2415-87-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...